Assay Details
| Gene Symbol: DNAJB8-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037890.1 | 2 - 3 | 347 | 82 | ||
| GenBank mRNA | AK097457.1 | - | 2 - 3 | 347 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DnaJ heat shock protein family (Hsp40) member B8 | DNAJB8 |
CT156, DJ6 |
Anti-sense | Chr. 3 - 128462432 - 128467248 on NCBI Build GRCh38 | |
| Downstream | GATA binding protein 2 | GATA2 |
DCML, IMD21, MONOMAC, NFE1B |
7.1kb | Anti-sense | Chr. 3 - 128479422 - 128493187 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000674.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 110822 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 302643 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 327857 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 342663 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 46539 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3566559 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 19818 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P00808597 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P00808597 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3566559 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3566559 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P00808597 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00808597 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 142830 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 7560 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00808591 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00808597 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16399840 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00808597 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00808591 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1562740_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012102.hg.1 | TC03000674.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012102.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012102.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012103.hg.1 | TC03000674.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012103.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012103.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012104.hg.1 | TC03000674.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012104.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012104.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162014.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162014.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162014.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162014.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162014.hg.1 | TC0300008723.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300162016.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162016.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162016.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162016.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162016.hg.1 | TC0300008723.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300162024.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162024.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162024.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162024.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162024.hg.1 | TC0300008723.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300162020.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162020.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162020.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162020.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162020.hg.1 | TC0300008723.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300162022.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162022.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162022.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162022.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162022.hg.1 | TC0300008723.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300162023.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162023.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162023.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162023.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162023.hg.1 | TC0300008723.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300162019.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162019.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162019.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162019.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162019.hg.1 | TC0300008723.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300162015.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162015.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162015.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162015.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162015.hg.1 | TC0300008723.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300162017.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162017.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162017.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162017.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162017.hg.1 | TC0300008723.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300162025.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162025.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162025.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300162025.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300162025.hg.1 | TC0300008723.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | ENST00000471626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008723.hg.1 | NR_037890.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1562740_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1562740_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562740_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562740_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562740_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562740_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562740_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562740_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562740_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562740_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562740_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562740_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562740_at4 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A023161 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A023161 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 625107 | - | - |

DNAJB8-AS1
Genomic Map