Assay Details
| Gene Symbol: MAVS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206491.1 | NP_001193420.1 | 5 - 6 | 1155 | 71 | |
| NM_020746.4 | NP_065797.2 | 6 - 7 | 1330 | 71 | ||
| NR_037921.1 | 5 - 6 | 1157 | 71 | |||
| GenBank mRNA | AB033097.1 | - | 5 - 6 | 1236 | 71 | |
| AB232371.1 | - | 5 - 6 | 1230 | 71 | ||
| AK291785.1 | - | 5 - 6 | 1114 | 71 | ||
| AK296897.1 | - | 5 - 6 | 1122 | 71 | ||
| BC044952.1 | - | 6 - 7 | 1264 | 5751684 | 71 | |
| BX649084.1 | - | 5 - 6 | 1116 | 71 | ||
| DQ167126.1 | - | 5 - 6 | 1158 | 71 | ||
| DQ174270.1 | - | 5 - 6 | 1158 | 71 | ||
| DQ181928.1 | - | 5 - 6 | 1158 | 71 | ||
| EF467323.1 | - | 4 - 5 | 983 | 71 | ||
| EF467324.1 | - | 4 - 5 | 985 | 71 | ||
| KC415005.1 | - | 5 - 6 | 1158 | 71 | ||
More Information
Set Membership: |
|

MAVS
Genomic Map