Assay Details
| Gene Symbol: RHOU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037962.1 | 1 - 2 | 387 | 89 | ||
| GenBank mRNA | DA321957.1 | - | 1 - 2 | 388 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105373132 | LOC105373132 | Sense | Chr. 1 - 228646284 - 228650007 on NCBI Build GRCh38 | ||
| Downstream |
RAB4A, member RAS oncogene family |
RAB4A |
HRES-1, HRES-1/RAB4, HRES1, RAB4 |
533.3kb | Sense | Chr. 1 - 229271062 - 229305894 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

RHOU
Genomic Map