Assay Details
| Gene Symbol: TTC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271420.1 | NP_001258349.1 | 2 - 3 | 916 | 61 | |
| GenBank mRNA | AK055780.1 | - | 2 - 3 | 472 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger SWIM-type containing 7 | ZSWIM7 | SWS1 | -426.0 | Anti-sense | Chr. 17 - 15976560 - 15999692 on NCBI Build GRCh38 |
| Downstream | nuclear receptor corepressor 1 | NCOR1 |
N-CoR, N-CoR1, PPP1R109, TRAC1, hN-CoR |
28.1kb | Anti-sense | Chr. 17 - 16030094 - 16216746 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK055780.1,NM_001271420.1 transcripts.
Set Membership: |
|

TTC19
Genomic Map