Assay Details
| Gene Symbol: NOC2LP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002826.2 | 1 | 2451 | 60 | ||
| GenBank mRNA | BC060851.1 | - | 1 | 2444 | 60 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1120 | LINC01120 | -33.9kb | Anti-sense | Chr. 2 - 131402901 - 131408228 on NCBI Build GRCh38 | |
| Downstream | mitotic spindle organizing protein 2A | MZT2A |
FAM128A, MOZART2A |
24.8kb | Sense | Chr. 2 - 131469724 - 131492719 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002352.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21601343 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01480707 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01480713 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01480704 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15327856 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21601242 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21601243 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21601326 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21601246 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21601247 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15327925 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01480714 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15327865 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21601245 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 58689 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 91957 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 223381 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 3545 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P93012 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 31993 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P336015 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01480712 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15327932 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15327925 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01480714 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01480716 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21601325 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01480713 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21601343 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21601326 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01480714 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15327925 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21601326 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01480713 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21601325 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01480712 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01480716 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15327932 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21601343 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P93012 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P93012 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01480703 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01480718 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01480706 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01480713 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15327921 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15327917 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01480710 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15327919 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01480714 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01480708 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_002826_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8055283 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8055282 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8055281 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR02037197.hg.1 | TC02002352.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02037197.hg.1 | NR_002826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02037198.hg.1 | TC02002352.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02037198.hg.1 | ENST00000434348.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02037198.hg.1 | NR_002826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009285.hg.1 | ENST00000450578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009285.hg.1 | ENST00000450578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009285.hg.1 | ENST00000452600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009285.hg.1 | ENST00000452600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200204492.hg.1 | ENST00000407594.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200204492.hg.1 | NR_002826.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200204492.hg.1 | ENST00000407594.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200204492.hg.1 | NR_002826.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200204492.hg.1 | TC0200014317.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200204491.hg.1 | NR_002826.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200204491.hg.1 | NR_002826.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200204491.hg.1 | TC0200014317.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200204493.hg.1 | NR_002826.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200204493.hg.1 | NR_002826.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200204493.hg.1 | TC0200014317.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200009285.hg.1 | ENST00000450578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009285.hg.1 | ENST00000450578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009285.hg.1 | ENST00000452600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009285.hg.1 | ENST00000452600.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014379 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014378 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014378 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014379 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2137499 | BC060851.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1651341 | NR_002826.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1651341 | NR_002826.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2137499 | BC060851.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2137499 | BC060851.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2137499 | BC060851.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1651341 | NR_002826.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2137499 | BC060851.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1651341 | NR_002826.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2137499 | BC060851.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2137499 | BC060851.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2137499 | BC060851.1 | human_transcriptome |

NOC2LP2
Genomic Map