Assay Details
| Gene Symbol: SNORD20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002908.1 | 1 | 43 | 80 | ||
| GenBank mRNA | Z34290.1 | - | 1 | 43 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, H/ACA box 75 |
SNORA75 |
SNORA75A, U23 |
-508.0 | Sense | Chr. 2 - 231455800 - 231455936 on NCBI Build GRCh38 |
| Downstream |
small nucleolar RNA, C/D box 82 |
SNORD82 |
RNU82, U82, Z25 |
3.8kb | Sense | Chr. 2 - 231460368 - 231460442 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002861.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3913 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01959201 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085764 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01959216 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085761 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085741 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085766 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085771 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01959208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085760 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01959197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085745 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01959212 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085768 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085765 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085748 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15392236 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085750 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085745 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085765 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01959201 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085748 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085750 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085771 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085766 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01959212 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01959208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085741 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085761 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085764 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01959197 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01959216 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085768 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15392236 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22085760 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959197 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959199 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959238 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15392238 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15392251 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15392246 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959212 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15392244 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959252 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959231 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15392236 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959240 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959253 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959247 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959237 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959229 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959205 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959215 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959236 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959218 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959202 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959222 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959233 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959235 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959194 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959249 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959246 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959243 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959251 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959245 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959227 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959209 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P01959207 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_002908_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8059711 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8059710 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016020.hg.1 | ENST00000384550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016020.hg.1 | ENST00000626841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016020.hg.1 | ENST00000626841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016020.hg.1 | ENST00000626841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016020.hg.1 | NR_002908.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02046203.hg.1 | TC02002861.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02046203.hg.1 | ENST00000384550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02046203.hg.1 | NR_002908.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016020.hg.1 | ENST00000384550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016020.hg.1 | ENST00000626841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016020.hg.1 | ENST00000626841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016020.hg.1 | ENST00000626841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016020.hg.1 | NR_002908.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217791.hg.1 | ENST00000384550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217791.hg.1 | ENST00000626841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217791.hg.1 | NR_002908.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217791.hg.1 | ENST00000384550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217791.hg.1 | ENST00000626841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217791.hg.1 | NR_002908.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217791.hg.1 | TC0200016020.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3241738 | NR_002908.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3241738 | Z34290.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3241738 | NR_002908.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3241738 | Z34290.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3241738 | NR_002908.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3241738 | Z34290.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3241738 | NR_002908.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3241738 | Z34290.1 | human_transcriptome |

SNORD20
Genomic Map