Assay Details
| Gene Symbol: LOC100128573 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024491.1 | 1 | 150 | 78 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rho/Rac guanine nucleotide exchange factor 18 | ARHGEF18 | P114-RhoGEF | Anti-sense | Chr. 19 - 7348901 - 7476990 on NCBI Build GRCh38 | |
| Downstream | peroxisomal biogenesis factor 11 gamma | PEX11G | 3.5kb | Sense | Chr. 19 - 7476870 - 7494977 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001112.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3276748 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4618 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4617 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 22668 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3276748 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3276748 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17104203 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31485213 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872896 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31485219 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872899 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872888 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872889 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872892 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872891 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872890 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872887 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31485222 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31485211 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872895 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872897 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872894 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17104204 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17104206 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872893 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872898 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31485211 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17104206 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10872890 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10872898 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17104204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10872894 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10872896 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10872889 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10872887 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10872892 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17104203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10872893 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872890 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872893 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872888 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872894 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872887 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17104203 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872895 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872896 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872898 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17104204 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31485213 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17104206 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31485219 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872899 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872892 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31485211 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872897 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872891 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31485222 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10872889 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10872887 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17104203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10872898 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17104204 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10872892 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10872893 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10872889 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10872894 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31485211 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10872896 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17104206 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10872890 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17104215 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17104206 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17104202 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10872897 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10872890 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10872927 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10872892 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10872894 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10872925 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10872931 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10872929 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17104216 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10872888 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17104204 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8025167 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8025168 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8025166 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8025142 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015288.hg.1 | NR_024491.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015288.hg.1 | TC19001112.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900176208.hg.1 | NR_024491.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176208.hg.1 | TC1900009478.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900176208.hg.1 | NR_024491.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010960 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010960 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3237079 | NR_024491.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3237079 | NR_024491.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237079 | NR_024491.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237079 | NR_024491.1 | human_transcriptome |

LOC100128573
Genomic Map