Assay Details
| Gene Symbol: ATP6V0CP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037141.1 | 1 | 53 | 82 | ||
| GenBank mRNA | BQ447003.1 | - | 1 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | peripherin 2 | PRPH2 |
AOFMD, AVMD, CACD2, DS, MDBS1, PRPH, RDS, RP7, TSPAN22, rd2 |
-4.6kb | Anti-sense | Chr. 6 - 42694509 - 42723024 on NCBI Build GRCh38 |
| Downstream | tubulin folding cofactor C | TBCC | CFC | 16.3kb | Anti-sense | Chr. 6 - 42744496 - 42746146 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000583.hg.1
Set Membership: |
|

ATP6V0CP3
Genomic Map