Assay Details
| Gene Symbol: ACTG1P17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036446.1 | 3 | 850 | 87 | ||
| GenBank mRNA | BC020234.1 | - | 3 | 850 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | epididymal protein pseudogene | LOC338963 | -11.9kb | Sense | Chr. 15 - 82710471 - 82713993 on NCBI Build GRCh38 | |
| Downstream | small nucleolar RNA host gene 21 | SNHG21 |
FSD2-AS1, SCARNA15HG |
23.1kb | Anti-sense | Chr. 15 - 82750564 - 82757206 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001758.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09792886 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09792836 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30294216 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09792828 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30294221 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09792831 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09792884 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09792882 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30294214 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30294212 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30294217 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30294206 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30294218 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09792830 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09792825 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09792876 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09792877 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30294259 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30294220 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30294211 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3297946 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 22544 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3297946 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3297946 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P314061 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7991018 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7991017 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1569368_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017238.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017238.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017238.hg.1 | TC15001758.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017237.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017237.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017237.hg.1 | TC15001758.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010247.hg.1 | ENST00000558391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010247.hg.1 | ENST00000558391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010247.hg.1 | ENST00000558391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010247.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010247.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010247.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010247.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010247.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010247.hg.1 | ENST00000561222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010247.hg.1 | ENST00000561222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010247.hg.1 | ENST00000561222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010247.hg.1 | ENST00000577648.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010247.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010247.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010247.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010247.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010247.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177081.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177081.hg.1 | ENST00000561222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177081.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177081.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177081.hg.1 | ENST00000561222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177081.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177081.hg.1 | TC1500010247.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500177075.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177075.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177075.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177075.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177075.hg.1 | TC1500010247.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500177076.hg.1 | ENST00000558391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177076.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177076.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177076.hg.1 | ENST00000558391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177076.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177076.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177076.hg.1 | TC1500010247.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500010247.hg.1 | ENST00000558391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010247.hg.1 | ENST00000558391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010247.hg.1 | ENST00000558391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010247.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010247.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010247.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010247.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010247.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010247.hg.1 | ENST00000561222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010247.hg.1 | ENST00000561222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010247.hg.1 | ENST00000561222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010247.hg.1 | ENST00000577648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010247.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010247.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010247.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010247.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010247.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177077.hg.1 | ENST00000558391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177077.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177077.hg.1 | ENST00000577648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177077.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177077.hg.1 | ENST00000558391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177077.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177077.hg.1 | ENST00000577648.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177077.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177077.hg.1 | TC1500010247.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500177078.hg.1 | ENST00000558391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177078.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177078.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177078.hg.1 | ENST00000558391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177078.hg.1 | ENST00000560958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500177078.hg.1 | NR_036446.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500177078.hg.1 | TC1500010247.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1569368_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1569368_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569368_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569368_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569368_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569368_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569368_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569368_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569368_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569368_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569368_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569368_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1569368_at4 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052479 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A052479 | - | - |

ACTG1P17
Genomic Map