Assay Details
| Gene Symbol: FOXN3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036500.1 | 2 | 852 | 74 | ||
| GenBank mRNA | BC067103.1 | - | 2 | 852 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tetratricopeptide repeat domain 8 | TTC8 |
BBS8, RP51 |
-541.2kb | Sense | Chr. 14 - 88824153 - 88877996 on NCBI Build GRCh38 |
| Downstream | FOXN3 antisense RNA 2 | FOXN3-AS2 | PRO1768 | 156.4kb | Sense | Chr. 14 - 89576216 - 89577477 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000544.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P306720 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 25299 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 12718 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P306720 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P306720 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P371708 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 227887_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 227885_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 229652_PM_s_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007925.hg.1 | ENST00000555562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007925.hg.1 | ENST00000555562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007925.hg.1 | ENST00000555562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007925.hg.1 | NR_036500.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007925.hg.1 | NR_036500.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007925.hg.1 | NR_036500.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006667.hg.1 | ENST00000555562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006667.hg.1 | NR_036500.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006667.hg.1 | TC14000544.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006666.hg.1 | ENST00000555562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006666.hg.1 | NR_036500.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14006666.hg.1 | TC14000544.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400156143.hg.1 | ENST00000555562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156143.hg.1 | NR_036500.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156143.hg.1 | ENST00000555562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156143.hg.1 | NR_036500.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156143.hg.1 | TC1400007925.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400156144.hg.1 | ENST00000555562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156144.hg.1 | NR_036500.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156144.hg.1 | ENST00000555562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156144.hg.1 | NR_036500.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156144.hg.1 | TC1400007925.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400156145.hg.1 | ENST00000555562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156145.hg.1 | NR_036500.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156145.hg.1 | ENST00000555562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156145.hg.1 | NR_036500.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156145.hg.1 | TC1400007925.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400007925.hg.1 | ENST00000555562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007925.hg.1 | ENST00000555562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007925.hg.1 | ENST00000555562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007925.hg.1 | NR_036500.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007925.hg.1 | NR_036500.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007925.hg.1 | NR_036500.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 227887_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 227885_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 229652_PM_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 229652_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 227885_at | - | - |
| Human Genome U133 Plus 2.0 Array | 227887_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227885_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227885_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229652_s_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229652_s_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227887_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229652_s_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229652_s_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229652_s_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229652_s_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227887_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227887_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227887_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227885_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227887_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229652_s_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227885_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227885_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227887_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227885_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229652_s_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227887_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227885_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229652_s_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227885_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227887_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227885_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227887_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227887_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227885_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229652_s_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227885_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 227887_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 229652_s_at3 | - | - |
| Human Genome U95 Set | 74995_at | - | - |
| Human Genome U95 Set | 74995_PM_at | - | - |
| Human Genome U95 Set | 78385_at | - | - |
| Human Genome U95 Set | 78385_PM_at | - | - |
| Human Genome U133 Set | 229652_PM_s_at | - | - |
| Human Genome U133 Set | 227887_at | - | - |
| Human Genome U133 Set | 229652_s_at | - | - |
| Human Genome U133 Set | 227887_PM_at | - | - |
| Human Genome U133 Set | 227885_PM_at | - | - |
| Human Genome U133 Set | 227885_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051683 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051683 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 163484 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1806892 | NR_036500.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1806892 | BC067103.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1806892 | NR_036500.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1806892 | BC067103.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1806892 | NR_036500.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1806892 | BC067103.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1806892 | NR_036500.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1806892 | BC067103.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1806892 | NR_036500.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1806892 | BC067103.1 | human_transcriptome |

FOXN3-AS1
Genomic Map