Assay Details
| Gene Symbol: NDUFB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199987.1 | NP_001186916.1 | 1 - 2 | 306 | 188 | |
| GenBank mRNA | AW160372.1 | - | 1 - 2 | 237 | 188 | |
More Information
Set Membership: |
|

NDUFB6
Genomic Map