Assay Details
| Gene Symbol: AP5S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204446.1 | NP_001191375.1 | 2 - 3 | 523 | 91 | |
| NM_001204447.1 | NP_001191376.1 | 2 - 3 | 371 | 91 | ||
| NM_018347.2 | NP_060817.1 | 2 - 3 | 427 | 91 | ||
| GenBank mRNA | AK002030.1 | - | 2 - 3 | 395 | 91 | |
| BC043344.1 | - | 2 - 3 | 364 | 5505030 | 91 | |
| DA129391.1 | - | 2 - 3 | 491 | 91 | ||
| DA253969.1 | - | 2 - 3 | 427 | 91 | ||
| DB190907.1 | - | 2 - 3 | 371 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000042.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000379567.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000379567.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000379567.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000379567.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000379567.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000379567.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000379567.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000379573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000379573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000379573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000379573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000379573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000379573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000379573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000379573.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000455742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000455742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000455742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000455742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000455742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000455742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_001204447.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_001204447.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_001204447.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_001204447.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_001204447.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_001204447.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_001204447.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_018347.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_018347.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_018347.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_018347.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_018347.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_018347.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_018347.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009884.hg.1 | NM_018347.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000723.hg.1 | ENST00000246041.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000723.hg.1 | ENST00000379567.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000723.hg.1 | ENST00000379573.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000723.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000723.hg.1 | NM_001204447.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000723.hg.1 | NM_018347.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000723.hg.1 | TC20000042.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000379567.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000379567.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000379567.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000379567.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000379567.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000379567.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000379567.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000379573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000379573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000379573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000379573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000379573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000379573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000379573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000379573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000455742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000455742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000455742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000455742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000455742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000455742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_001204447.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_001204447.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_001204447.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_001204447.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_001204447.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_001204447.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_001204447.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_018347.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_018347.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_018347.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_018347.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_018347.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_018347.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_018347.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009884.hg.1 | NM_018347.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000169257.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000169257.hg.1 | ENST00000379567.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000169257.hg.1 | ENST00000379573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000169257.hg.1 | ENST00000455742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000169257.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000169257.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000169257.hg.1 | NM_001204447.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000169257.hg.1 | NM_018347.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000169257.hg.1 | TC2000009884.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000169257.hg.1 | ENST00000246041.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000169257.hg.1 | ENST00000379567.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000169257.hg.1 | ENST00000379573.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000169257.hg.1 | ENST00000455742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000169257.hg.1 | ENST00000615891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000169257.hg.1 | NM_001204446.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000169257.hg.1 | NM_001204447.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000169257.hg.1 | NM_018347.2 | Refseq |

AP5S1
Genomic Map