Assay Details
| Gene Symbol: KIFAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204514.1 | NP_001191443.1 | 12 - 13 | 1724 | 96 | |
| NM_001204516.1 | NP_001191445.1 | 10 - 11 | 1428 | 96 | ||
| NM_001204517.1 | NP_001191446.1 | 11 - 12 | 2669 | 96 | ||
| NM_014970.3 | NP_055785.2 | 11 - 12 | 1560 | 96 | ||
| XM_005244970.1 | XP_005245027.1 | 11 - 12 | 1563 | 96 | ||
| XM_011509307.2 | XP_011507609.2 | 11 - 12 | 1563 | 96 | ||
| GenBank mRNA | AK026471.1 | - | 11 - 12 | 1339 | 96 | |
| AK122897.1 | - | 11 - 12 | 2671 | 96 | ||
| AK297368.1 | - | 10 - 11 | 1355 | 96 | ||
| AK301912.1 | - | 4 - 5 | 527 | 96 | ||
| AK303052.1 | - | 12 - 13 | 1681 | 96 | ||
| AK315586.1 | - | 11 - 12 | 1474 | 96 | ||
| BC028679.2 | - | 11 - 12 | 1517 | 4829354 | 96 | |
| BC048012.1 | - | 10 - 11 | 1472 | 96 | ||
| U59919.1 | - | 11 - 12 | 1477 | 96 | ||
More Information
Set Membership: |
|

KIFAP3
Genomic Map