Assay Details
| Gene Symbol: KIFAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204514.1 | NP_001191443.1 | 18 - 19 | 2419 | 104 | |
| NM_001204516.1 | NP_001191445.1 | 16 - 17 | 2123 | 104 | ||
| NM_001204517.1 | NP_001191446.1 | 17 - 18 | 3364 | 104 | ||
| NM_014970.3 | NP_055785.2 | 17 - 18 | 2255 | 104 | ||
| XM_005244970.1 | XP_005245027.1 | 17 - 18 | 2258 | 104 | ||
| XM_011509307.2 | XP_011507609.2 | 17 - 18 | 2258 | 104 | ||
| GenBank mRNA | AK026471.1 | - | 17 - 18 | 2034 | 104 | |
| AK122897.1 | - | 17 - 18 | 3366 | 104 | ||
| AK297368.1 | - | 16 - 17 | 2050 | 104 | ||
| AK301912.1 | - | 10 - 11 | 1222 | 104 | ||
| AK303052.1 | - | 18 - 19 | 2376 | 104 | ||
| AK315586.1 | - | 17 - 18 | 2169 | 104 | ||
| BC028679.2 | - | 17 - 18 | 2212 | 4829354 | 104 | |
| BC048012.1 | - | 16 - 17 | 2167 | 104 | ||
| U59919.1 | - | 17 - 18 | 2172 | 104 | ||
More Information
Set Membership: |
|

KIFAP3
Genomic Map