Assay Details
| Gene Symbol: KIFAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204514.1 | NP_001191443.1 | 5 - 6 | 815 | 123 | |
| NM_001204516.1 | NP_001191445.1 | 3 - 4 | 519 | 123 | ||
| NM_001204517.1 | NP_001191446.1 | 4 - 5 | 1760 | 123 | ||
| NM_014970.3 | NP_055785.2 | 4 - 5 | 651 | 123 | ||
| XM_005244970.1 | XP_005245027.1 | 4 - 5 | 654 | 123 | ||
| XM_011509307.2 | XP_011507609.2 | 4 - 5 | 654 | 123 | ||
| GenBank mRNA | AK026471.1 | - | 4 - 5 | 430 | 123 | |
| AK122897.1 | - | 4 - 5 | 1762 | 123 | ||
| AK297368.1 | - | 3 - 4 | 446 | 123 | ||
| AK303052.1 | - | 5 - 6 | 772 | 123 | ||
| AK315586.1 | - | 4 - 5 | 565 | 123 | ||
| BC028679.2 | - | 4 - 5 | 608 | 4829354 | 123 | |
| BC048012.1 | - | 3 - 4 | 563 | 123 | ||
| U59919.1 | - | 4 - 5 | 568 | 123 | ||
More Information
Set Membership: |
|

KIFAP3
Genomic Map