Assay Details
| Gene Symbol: KIFAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204514.1 | NP_001191443.1 | 8 - 9 | 1180 | 103 | |
| NM_001204516.1 | NP_001191445.1 | 6 - 7 | 884 | 103 | ||
| NM_001204517.1 | NP_001191446.1 | 7 - 8 | 2125 | 103 | ||
| NM_014970.3 | NP_055785.2 | 7 - 8 | 1016 | 103 | ||
| XM_005244970.1 | XP_005245027.1 | 7 - 8 | 1019 | 103 | ||
| XM_011509307.2 | XP_011507609.2 | 7 - 8 | 1019 | 103 | ||
| GenBank mRNA | AK026471.1 | - | 7 - 8 | 795 | 103 | |
| AK122897.1 | - | 7 - 8 | 2127 | 103 | ||
| AK297368.1 | - | 6 - 7 | 811 | 103 | ||
| AK303052.1 | - | 8 - 9 | 1137 | 103 | ||
| AK315586.1 | - | 7 - 8 | 930 | 103 | ||
| BC028679.2 | - | 7 - 8 | 973 | 4829354 | 103 | |
| BC048012.1 | - | 6 - 7 | 928 | 103 | ||
| U59919.1 | - | 7 - 8 | 933 | 103 | ||
More Information
Set Membership: |
|

KIFAP3
Genomic Map