| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01017215.hg.1 |
TC01001093.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01017215.hg.1 |
ENST00000330165.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01017215.hg.1 |
ENST00000369307.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01017215.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000484825.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000484825.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000484825.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000484825.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000484825.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000498663.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000498663.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000498663.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000498663.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000498663.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000498663.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000498663.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000632555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000632555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000632555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000632555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000632555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000632555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000632555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000632555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000634161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000634161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000634161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000634161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
ENST00000634161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000484825.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000484825.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000484825.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000484825.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000484825.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000498663.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000498663.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000498663.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000498663.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000498663.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000498663.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000498663.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000632555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000632555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000632555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000632555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000632555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000632555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000632555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000632555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000634161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000634161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000634161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000634161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
ENST00000634161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018477.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100242814.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242814.hg.1 |
ENST00000484825.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242814.hg.1 |
ENST00000498663.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242814.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242814.hg.1 |
ENST00000632555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242814.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100242814.hg.1 |
ENST00000369307.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242814.hg.1 |
ENST00000484825.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242814.hg.1 |
ENST00000498663.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242814.hg.1 |
ENST00000583313.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242814.hg.1 |
ENST00000632555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100242814.hg.1 |
NM_005105.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100242814.hg.1 |
TC0100018477.hg.1 |
Clariom D TC |