Assay Details
| Gene Symbol: TUSC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006765.3 | NP_006756.2 | 8 - 9 | 1284 | 65 | |
| NM_178234.2 | NP_839952.1 | 8 - 9 | 1284 | 65 | ||
| XM_011544651.2 | XP_011542953.1 | 7 - 8 | 827 | 65 | ||
| XM_011544652.2 | XP_011542954.1 | 7 - 8 | 827 | 65 | ||
| XM_017013860.1 | XP_016869349.1 | 7 - 8 | 827 | 65 | ||
| XM_017013861.1 | XP_016869350.1 | 7 - 8 | 827 | 65 | ||
| GenBank mRNA | AY562998.1 | - | 1 - 2 | 43 | 65 | |
| AY562999.1 | - | 1 - 2 | 43 | 65 | ||
| BC010370.1 | - | 8 - 9 | 1152 | 4334284 | 65 | |
| BT020002.1 | - | 8 - 9 | 940 | 65 | ||
| U42349.1 | - | 8 - 9 | 1097 | 65 | ||
More Information
Set Membership: |
|

TUSC3
Genomic Map