Assay Details
| Gene Symbol: INPP5E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318502.1 | NP_001305431.1 | 4 - 5 | 1558 | 83 | |
| NM_019892.5 | NP_063945.2 | 4 - 5 | 1558 | 83 | ||
| XM_017014926.1 | XP_016870415.1 | 4 - 5 | 1597 | 83 | ||
| GenBank mRNA | AF187891.1 | - | 4 - 5 | 1259 | 83 | |
| AL832846.1 | - | 4 - 5 | 1559 | 83 | ||
| BC028032.1 | - | 4 - 5 | 1540 | 5242186 | 83 | |
More Information
Set Membership: |
|

INPP5E
Genomic Map