Assay Details
| Gene Symbol: INPP5E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318502.1 | NP_001305431.1 | 6 - 7 | 1790 | 70 | |
| NM_019892.5 | NP_063945.2 | 6 - 7 | 1793 | 70 | ||
| XM_017014926.1 | XP_016870415.1 | 6 - 7 | 1832 | 70 | ||
| GenBank mRNA | AF187891.1 | - | 6 - 7 | 1494 | 70 | |
| AL832846.1 | - | 6 - 7 | 1791 | 70 | ||
| BC028032.1 | - | 6 - 7 | 1775 | 5242186 | 70 | |
More Information
Set Membership: |
|

INPP5E
Genomic Map