Assay Details
| Gene Symbol: CCNE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057749.2 | NP_477097.1 | 3 - 4 | 211 | 71 | |
| XM_011517366.2 | XP_011515668.1 | 2 - 3 | 536 | 71 | ||
| XM_017013957.1 | XP_016869446.1 | 2 - 3 | 536 | 71 | ||
| XM_017013958.1 | XP_016869447.1 | 3 - 4 | 353 | 71 | ||
| XM_017013959.1 | XP_016869448.1 | 3 - 4 | 455 | 71 | ||
| GenBank mRNA | AF091433.1 | - | 2 - 3 | 144 | 71 | |
| AF102778.1 | - | 1 - 2 | 107 | 71 | ||
| AF106690.1 | - | 1 - 2 | 107 | 71 | ||
| AF112857.1 | - | 1 - 2 | 107 | 71 | ||
| BC020729.1 | - | 3 - 4 | 158 | 4774554 | 71 | |
| BG720611.1 | - | 3 - 4 | 216 | 71 | ||
| BJ990268.1 | - | 3 - 4 | 211 | 71 | ||
More Information
Set Membership: |
|

CCNE2
Genomic Map