Assay Details
| Gene Symbol: CCNE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057749.2 | NP_477097.1 | 6 - 7 | 561 | 91 | |
| XM_011517366.2 | XP_011515668.1 | 5 - 6 | 886 | 91 | ||
| XM_017013957.1 | XP_016869446.1 | 5 - 6 | 886 | 91 | ||
| XM_017013958.1 | XP_016869447.1 | 6 - 7 | 703 | 91 | ||
| XM_017013959.1 | XP_016869448.1 | 6 - 7 | 805 | 91 | ||
| GenBank mRNA | AF091433.1 | - | 5 - 6 | 494 | 91 | |
| AF102778.1 | - | 4 - 5 | 457 | 91 | ||
| AF106690.1 | - | 4 - 5 | 457 | 91 | ||
| BC007015.1 | - | 4 - 5 | 296 | 3997513 | 91 | |
| BC020729.1 | - | 6 - 7 | 508 | 4774554 | 91 | |
More Information
Set Membership: |
|

CCNE2
Genomic Map