Assay Details
| Gene Symbol: TCTEX1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152773.4 | NP_689986.2 | 2 - 3 | 391 | 95 | |
| GenBank mRNA | BC021177.2 | - | 2 - 3 | 371 | 4830500 | 95 |
| BM548106.1 | - | 2 - 3 | 416 | 95 | ||
| CK818899.1 | - | 2 - 3 | 385 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03003401.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03038207.hg.1 | TC03003401.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038207.hg.1 | ENST00000325318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038207.hg.1 | ENST00000545438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038207.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038202.hg.1 | TC03003401.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038202.hg.1 | ENST00000325318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038202.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038211.hg.1 | TC03003401.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038211.hg.1 | ENST00000325318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038211.hg.1 | ENST00000545438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038211.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000465757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000465757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000465757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000465757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000491186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000491186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | ENST00000491186.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014094.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038209.hg.1 | TC03003401.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038209.hg.1 | ENST00000325318.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038209.hg.1 | ENST00000545438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03038209.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300210013.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210013.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210013.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300210013.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210013.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210013.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300210013.hg.1 | TC0300014094.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000465757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000465757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000465757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000465757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000491186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000491186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | ENST00000491186.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014094.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300210006.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210006.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210006.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210006.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300210006.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210006.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210006.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210006.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300210006.hg.1 | TC0300014094.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300210004.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210004.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210004.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210004.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300210004.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210004.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210004.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210004.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300210004.hg.1 | TC0300014094.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300210011.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210011.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210011.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210011.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300210011.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210011.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210011.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210011.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300210011.hg.1 | TC0300014094.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300210010.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210010.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210010.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210010.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300210010.hg.1 | ENST00000325318.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210010.hg.1 | ENST00000426563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210010.hg.1 | ENST00000446494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300210010.hg.1 | NM_152773.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300210010.hg.1 | TC0300014094.hg.1 | Clariom D TC |

TCTEX1D2
Genomic Map