Assay Details
Gene Symbol: MTRNR2L8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190702.1 | NP_001177631.1 | 1 | 1046 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001411.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
ArrayStar
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000009588.hg.1 | ENST00000543500.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009588.hg.1 | ENST00000543500.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009588.hg.1 | ENST00000543500.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009588.hg.1 | ENST00000543500.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009588.hg.1 | NM_001190472.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009588.hg.1 | NM_001190472.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009588.hg.1 | NM_001190472.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11017749.hg.1 | ENST00000536684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11017749.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11017749.hg.1 | TC11001411.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700007296.hg.1 | ENST00000540040.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007296.hg.1 | ENST00000540040.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007296.hg.1 | ENST00000540040.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007296.hg.1 | NM_001190452.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007296.hg.1 | NM_001190452.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007296.hg.1 | NM_001190452.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007183.hg.1 | ENST00000582078.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007183.hg.1 | ENST00000582078.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007183.hg.1 | ENST00000607661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007183.hg.1 | ENST00000607661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11017747.hg.1 | ENST00000536684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11017747.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11017747.hg.1 | TC11001411.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0400011114.hg.1 | ENST00000510231.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009462.hg.1 | ENST00000604952.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009462.hg.1 | ENST00000604952.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009462.hg.1 | ENST00000604952.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009462.hg.1 | ENST00000604952.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009462.hg.1 | ENST00000604952.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010087.hg.1 | NR_039705.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11017748.hg.1 | ENST00000536684.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11017748.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11017748.hg.1 | NR_039705.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11017748.hg.1 | TC11001411.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008379.hg.1 | ENST00000622562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008379.hg.1 | NM_001190706.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008379.hg.1 | NM_001190706.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008379.hg.1 | NM_001190706.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100176455.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100176455.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100176455.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100176455.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100176455.hg.1 | TC1100010087.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100176456.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100176456.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100176456.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100176456.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100176456.hg.1 | TC1100010087.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400011114.hg.1 | ENST00000510231.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007296.hg.1 | ENST00000540040.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007296.hg.1 | ENST00000540040.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007296.hg.1 | ENST00000540040.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007296.hg.1 | NM_001190452.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007296.hg.1 | NM_001190452.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007296.hg.1 | NM_001190452.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009588.hg.1 | ENST00000543500.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009588.hg.1 | ENST00000543500.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009588.hg.1 | ENST00000543500.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009588.hg.1 | ENST00000543500.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009588.hg.1 | NM_001190472.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009588.hg.1 | NM_001190472.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009588.hg.1 | NM_001190472.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011792.hg.1 | ENST00000600213.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007183.hg.1 | ENST00000582078.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007183.hg.1 | ENST00000582078.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007183.hg.1 | ENST00000607661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007183.hg.1 | ENST00000607661.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100176457.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100176457.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100176457.hg.1 | NR_039705.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100176457.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100176457.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100176457.hg.1 | NR_039705.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100176457.hg.1 | TC1100010087.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010087.hg.1 | NR_039705.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100176458.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100176458.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100176458.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100176458.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100176458.hg.1 | TC1100010087.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600008379.hg.1 | ENST00000622562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008379.hg.1 | NM_001190706.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008379.hg.1 | NM_001190706.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008379.hg.1 | NM_001190706.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100176454.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100176454.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100176454.hg.1 | ENST00000536684.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100176454.hg.1 | NM_001190702.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100176454.hg.1 | TC1100010087.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700009462.hg.1 | ENST00000604952.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009462.hg.1 | ENST00000604952.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009462.hg.1 | ENST00000604952.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009462.hg.1 | ENST00000604952.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009462.hg.1 | ENST00000604952.1 | Ensembl |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044933 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044933 | - | - |
