Assay Details
| Gene Symbol: HSCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318314.1 | NP_001305243.1 | 2 - 3 | 392 | 74 | |
| NM_001318315.1 | NP_001305244.1 | 2 - 3 | 392 | 74 | ||
| NM_001318316.1 | NP_001305245.1 | 2 - 3 | 141 | 74 | ||
| NM_172002.4 | NP_741999.3 | 2 - 3 | 392 | 74 | ||
| XM_005261359.3 | XP_005261416.1 | 2 - 3 | 393 | 74 | ||
| XM_017028619.1 | XP_016884108.1 | 2 - 3 | 358 | 74 | ||
| XM_017028620.1 | XP_016884109.1 | 2 - 3 | 196 | 74 | ||
| GenBank mRNA | AY191719.1 | - | 2 - 3 | 342 | 74 | |
| BC000004.1 | - | 2 - 3 | 401 | 74 | ||
| BC065569.1 | - | 2 - 3 | 365 | 6171959 | 74 | |
| BQ054405.1 | - | 2 - 3 | 369 | 74 | ||
| CR456462.1 | - | 2 - 3 | 360 | 74 | ||
| CR979598.1 | - | 2 - 3 | 341 | 74 | ||
| DW414316.1 | - | 2 - 3 | 157 | 74 | ||
| HY057929.1 | - | 2 - 3 | 141 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001318316.1, XM_017028619.1, XM_017028620.1 transcripts.
Set Membership: |
|

HSCB
Genomic Map