Assay Details
| Gene Symbol: GJD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020660.2 | NP_065711.1 | 2 | 261 | 128 | |
| XM_017022438.1 | XP_016877927.1 | 2 | 317 | 128 | ||
| GenBank mRNA | BC069339.1 | - | 2 | 168 | 7262267 | 128 |
| BC112110.1 | - | 2 | 261 | 8327578 | 128 | |
| BC112114.1 | - | 2 | 261 | 8327582 | 128 | |
| BC143788.1 | - | 2 | 261 | 9052302 | 128 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017022438.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

GJD2
Genomic Map