Assay Details
| Gene Symbol: PANX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160300.1 | NP_001153772.1 | 1 | 200 | 68 | |
| NM_052839.3 | NP_443071.2 | 1 | 200 | 68 | ||
| GenBank mRNA | AF398510.1 | - | 1 | 170 | 68 | |
| AK096019.1 | - | 1 | 184 | 68 | ||
| AK299910.1 | - | 1 | 200 | 68 | ||
| DA116770.1 | - | 1 | 202 | 68 | ||
| DA127683.1 | - | 1 | 200 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000419.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22007954.hg.1 | ENST00000159647.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007954.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007954.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007954.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007954.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007954.hg.1 | TC22000419.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007955.hg.1 | ENST00000159647.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007955.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007955.hg.1 | ENST00000401643.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007955.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007955.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007955.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22007955.hg.1 | TC22000419.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200155278.hg.1 | TC2200007789.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200155278.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155278.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155278.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155278.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155278.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155278.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155278.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155278.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155278.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155278.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155278.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155278.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155277.hg.1 | TC2200007789.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200155277.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155277.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155277.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155277.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155277.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155277.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155277.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200155277.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155277.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200155277.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000159647.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000395842.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | ENST00000402472.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_001160300.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NM_052839.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007789.hg.1 | NR_027691.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 105746 | - | - |

PANX2
Genomic Map