Assay Details
| Gene Symbol: CREB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004379.4 | NP_004370.1 | 7 - 8 | 1092 | 69 | |
| NM_134442.4 | NP_604391.1 | 8 - 9 | 1134 | 69 | ||
| NR_135473.1 | 8 - 9 | 1134 | 69 | |||
| XM_011510645.1 | XP_011508947.1 | 8 - 9 | 948 | 69 | ||
| XM_011510647.2 | XP_011508949.1 | 7 - 8 | 949 | 69 | ||
| XM_011510649.2 | XP_011508951.1 | 7 - 8 | 945 | 69 | ||
| XM_011510650.2 | XP_011508952.1 | 7 - 8 | 1044 | 69 | ||
| XM_011510651.2 | XP_011508953.1 | 6 - 7 | 861 | 69 | ||
| XM_017003400.1 | XP_016858889.1 | 6 - 7 | 948 | 69 | ||
| XM_017003401.1 | XP_016858890.1 | 7 - 8 | 806 | 69 | ||
| GenBank mRNA | AK312747.1 | - | 9 - 10 | 1029 | 69 | |
| AK315226.1 | - | 7 - 8 | 883 | 69 | ||
| BC010636.1 | - | 8 - 9 | 977 | 3872792 | 69 | |
| BC095407.1 | - | 7 - 8 | 1036 | 30344522 | 69 | |
| BT019509.1 | - | 7 - 8 | 883 | 69 | ||
| CR407617.1 | - | 6 - 7 | 841 | 69 | ||
| M27691.1 | - | 7 - 8 | 957 | 69 | ||
| M34356.1 | - | 7 - 8 | 883 | 69 | ||
| X55545.1 | - | 1026 | 69 | |||
More Information
Set Membership: |
|

CREB1
Genomic Map