Assay Details
| Gene Symbol: CALR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004343.3 | NP_004334.1 | 8 - 9 | 1132 | 66 | |
| GenBank mRNA | AB451408.1 | - | 8 - 9 | 1052 | 66 | |
| AK130190.1 | - | 3 - 4 | 428 | 66 | ||
| AK223060.1 | - | 8 - 9 | 1020 | 66 | ||
| AK295230.1 | - | 7 - 8 | 1031 | 66 | ||
| AK304492.1 | - | 7 - 8 | 1079 | 66 | ||
| AY047586.1 | - | 8 - 9 | 1105 | 66 | ||
| BC002500.2 | - | 8 - 9 | 1085 | 3050717 | 66 | |
| BC007911.1 | - | 8 - 9 | 1122 | 4299303 | 66 | |
| BC020493.1 | - | 8 - 9 | 1114 | 3898550 | 66 | |
| BC107102.2 | - | 1 - 2 | 66 | |||
| BT007448.1 | - | 8 - 9 | 1052 | 66 | ||
| CR457070.1 | - | 8 - 9 | 1052 | 66 | ||
| FJ224311.1 | - | 8 - 9 | 1118 | 66 | ||
| M32294.1 | - | 8 - 9 | 1118 | 66 | ||
| M84739.1 | - | 8 - 9 | 1160 | 66 | ||
More Information
Set Membership: |
|

CALR
Genomic Map