Assay Details
| Gene Symbol: CD8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145873.1 | NP_001139345.1 | 7 - 8 | 1666 | 89 | |
| NM_001768.6 | NP_001759.3 | 4 - 5 | 1524 | 89 | ||
| NR_027353.1 | 5 - 6 | 1110 | 89 | |||
| GenBank mRNA | AK097942.1 | - | 5 - 6 | 709 | 89 | |
| AK124156.1 | - | 7 - 8 | 1666 | 89 | ||
| AK300089.1 | - | 5 - 6 | 1076 | 89 | ||
| AY039664.1 | - | 4 - 5 | 814 | 89 | ||
| BC025715.1 | - | 4 - 5 | 640 | 5227906 | 89 | |
| M12824.1 | - | 4 - 5 | 722 | 89 | ||
| M12828.1 | - | 4 - 5 | 700 | 89 | ||
More Information
Set Membership: |
|

CD8A
Genomic Map