Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033025.hg.1 |
TC02002059.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033025.hg.1 |
ENST00000283635.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033025.hg.1 |
ENST00000352580.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033025.hg.1 |
ENST00000409511.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033025.hg.1 |
ENST00000442577.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033025.hg.1 |
ENST00000456996.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033025.hg.1 |
NM_001145873.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033025.hg.1 |
NM_001768.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033025.hg.1 |
NM_171827.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033025.hg.1 |
NR_027353.1 |
Refseq |