Assay Details
| Gene Symbol: CD8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001768.6 | NP_001759.3 | 1 | 516 | 60 | |
| NM_171827.3 | NP_741969.1 | 1 | 516 | 60 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001768.6,NM_171827.3 transcripts.
This assay is in the 5'UTR region of the NM_001768.6,NM_171827.3 transcripts.
Set Membership: |
|

CD8A
Genomic Map