Assay Details
| Gene Symbol: CD8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001768.6 | NP_001759.3 | 1 | 621 | 114 | |
| NM_171827.3 | NP_741969.1 | 1 | 621 | 114 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002059.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001768.6,NM_171827.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
T cell mediated immunity
immune response
cell surface receptor signaling pathway
transmembrane receptor protein tyrosine kinase signaling pathway
antigen processing and presentation
T cell activation
cytotoxic T cell differentiation
regulation of immune response
positive regulation of calcium-mediated signaling
defense response to virus
protein binding
coreceptor activity
protein kinase binding
MHC class I protein binding
protein homodimerization activity
T cell mediated immunity
immune response
cell surface receptor signaling pathway
transmembrane receptor protein tyrosine kinase signaling pathway
antigen processing and presentation
T cell activation
cytotoxic T cell differentiation
regulation of immune response
positive regulation of calcium-mediated signaling
defense response to virus
protein binding
coreceptor activity
protein kinase binding
MHC class I protein binding
protein homodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000352580.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000352580.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000352580.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000352580.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000352580.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000352580.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000352580.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000409781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000409781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000409781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000409781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | ENST00000409781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033045.hg.1 | TC02002059.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033045.hg.1 | ENST00000283635.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033045.hg.1 | ENST00000409511.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033045.hg.1 | ENST00000456996.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033045.hg.1 | ENST00000538832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033045.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033045.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033045.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02033045.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197408.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197408.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197408.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197408.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197408.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197408.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197408.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197408.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200197408.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197408.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197408.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197408.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200197408.hg.1 | TC0200013323.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000283635.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000352580.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000352580.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000352580.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000352580.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000352580.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000352580.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000352580.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000409511.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000409781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000409781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000409781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000409781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | ENST00000409781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001145873.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_001768.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NM_171827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NR_027353.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013323.hg.1 | NR_027353.1 | Refseq |

CD8A
Genomic Map