Assay Details
| Gene Symbol: GPR183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004951.4 | NP_004942.1 | 2 | 618 | 93 | |
| XM_017020405.1 | XP_016875894.1 | 2 | 561 | 93 | ||
| GenBank mRNA | AK223390.1 | - | 2 | 522 | 93 | |
| AK292091.1 | - | 2 | 542 | 93 | ||
| AK313443.1 | - | 2 | 618 | 93 | ||
| BC020752.1 | - | 2 | 522 | 4722289 | 93 | |
| L08177.1 | - | 1 | 477 | 93 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

GPR183
Genomic Map