Assay Details
| Gene Symbol: CD8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145873.1 | NP_001139345.1 | 9 | 2674 | 74 | |
| NM_001768.6 | NP_001759.3 | 6 | 2532 | 74 | ||
| NM_171827.3 | NP_741969.1 | 5 | 2421 | 74 | ||
| NR_027353.1 | 7 | 2118 | 74 | |||
| GenBank mRNA | AK097942.1 | - | 7 | 1716 | 74 | |
| AK124156.1 | - | 9 | 2674 | 74 | ||
| BC025715.1 | - | 6 | 1647 | 5227906 | 74 | |
| M12824.1 | - | 6 | 1730 | 74 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CD8A
Genomic Map