Assay Details
| Gene Symbol: MYO18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078471.3 | NP_510880.2 | 39 - 40 | 6006 | 78 | |
| GenBank mRNA | AB177858.1 | - | 38 - 39 | 5825 | 78 | |
| D86970.2 | - | 39 - 40 | 6029 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001315.hg.1
Set Membership: |
|

MYO18A
Genomic Map