Assay Details
| Gene Symbol: GZMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004131.4 | NP_004122.2 | 4 - 5 | 671 | 71 | |
| XM_011536685.2 | XP_011534987.1 | 4 - 5 | 655 | 71 | ||
| GenBank mRNA | AY232654.1 | - | 4 - 5 | 636 | 71 | |
| AY232655.1 | - | 3 - 4 | 473 | 71 | ||
| AY232656.1 | - | 2 - 3 | 326 | 71 | ||
| AY372494.1 | - | 4 - 5 | 712 | 71 | ||
| BC030195.1 | - | 4 - 5 | 627 | 5223876 | 71 | |
| CB528665.1 | - | 1 - 2 | 71 | |||
| J03189.1 | - | 4 - 5 | 644 | 71 | ||
| J04071.1 | - | 4 - 5 | 621 | 71 | ||
| M17016.1 | - | 4 - 5 | 612 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
protein processing
cytolysis
natural killer cell mediated cytotoxicity
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
serine-type endopeptidase activity
protein binding
serine-type peptidase activity
apoptotic process
protein processing
cytolysis
natural killer cell mediated cytotoxicity
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
serine-type endopeptidase activity
protein binding
serine-type peptidase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000382542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000382542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000382542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000382542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000382542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000382542.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000526004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000526004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000526004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000526004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000530830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000530830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000530830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000530830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000530830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000530830.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000532263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000532263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000532263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000532263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000532263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000554242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000554242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000554242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000554242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000554242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000554242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000616551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000616551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000616551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000616551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000616551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | ENST00000616551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008764.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164581.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164581.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164581.hg.1 | ENST00000382542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164581.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164581.hg.1 | ENST00000532263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164581.hg.1 | ENST00000554242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164581.hg.1 | ENST00000616551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164581.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164581.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164581.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164581.hg.1 | ENST00000382542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164581.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164581.hg.1 | ENST00000532263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164581.hg.1 | ENST00000554242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164581.hg.1 | ENST00000616551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164581.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164581.hg.1 | TC1400008764.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400164579.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164579.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164579.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164579.hg.1 | ENST00000616551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164579.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164579.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164579.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164579.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164579.hg.1 | ENST00000616551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164579.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164579.hg.1 | TC1400008764.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000382542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000382542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000382542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000382542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000382542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000382542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000526004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000526004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000526004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000526004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000530830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000530830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000530830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000530830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000530830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000530830.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000532263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000532263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000532263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000532263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000532263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000554242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000554242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000554242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000554242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000554242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000554242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000616551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000616551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000616551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000616551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000616551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | ENST00000616551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008764.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164580.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164580.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164580.hg.1 | ENST00000382542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164580.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164580.hg.1 | ENST00000532263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164580.hg.1 | ENST00000554242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164580.hg.1 | ENST00000616551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164580.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164580.hg.1 | ENST00000216341.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164580.hg.1 | ENST00000382540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164580.hg.1 | ENST00000382542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164580.hg.1 | ENST00000415355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164580.hg.1 | ENST00000532263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164580.hg.1 | ENST00000554242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164580.hg.1 | ENST00000616551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400164580.hg.1 | NM_004131.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400164580.hg.1 | TC1400008764.hg.1 | Clariom D TC |

GZMB
Genomic Map