Assay Details
| Gene Symbol: TOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014729.2 | NP_055544.1 | 1 - 2 | 331 | 65 | |
| GenBank mRNA | AB018351.1 | - | 1 - 2 | 333 | 65 | |
| AK000971.1 | - | 1 - 2 | 113 | 65 | ||
| AK302331.1 | - | 1 - 2 | 255 | 65 | ||
| BC016665.1 | - | 1 - 2 | 174 | 4342384 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK302331.1 transcripts.
Set Membership: |
|

TOX
Genomic Map