Assay Details
| Gene Symbol: GBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319179.1 | NP_001306108.1 | 11 - 12 | 1947 | 70 | |
| NM_001319180.1 | NP_001306109.1 | 10 - 11 | 2003 | 70 | ||
| NM_018284.2 | NP_060754.2 | 10 - 11 | 1861 | 70 | ||
| NR_135007.1 | 8 - 9 | 1367 | 70 | |||
| XM_005270743.2 | XP_005270800.1 | 11 - 12 | 1937 | 70 | ||
| XM_006710574.2 | XP_006710637.1 | 10 - 11 | 1794 | 70 | ||
| XM_011541233.1 | XP_011539535.1 | 9 - 10 | 1649 | 70 | ||
| XM_011541234.1 | XP_011539536.1 | 9 - 10 | 1632 | 70 | ||
| XM_017001013.1 | XP_016856502.1 | 9 - 10 | 1781 | 70 | ||
| XM_017001014.1 | XP_016856503.1 | 8 - 9 | 1529 | 70 | ||
| GenBank mRNA | AK001823.1 | - | 10 - 11 | 1975 | 70 | |
| AK309096.1 | - | 9 - 10 | 2501 | 70 | ||
| AL136680.1 | - | 10 - 11 | 1760 | 70 | ||
| BC063819.1 | - | 8 - 9 | 1279 | 70 | ||
| BC140837.1 | - | 10 - 11 | 1746 | 9021705 | 70 | |
| BC144650.1 | - | 11 - 12 | 1832 | 70 | ||
| CR936755.1 | - | 10 - 11 | 1862 | 70 | ||
| CR936758.1 | - | 10 - 11 | 1802 | 70 | ||
More Information
Set Membership: |
|

GBP3
Genomic Map