Assay Details
| Gene Symbol: LOC653653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027408.1 | 1 | 123 | 93 | ||
| GenBank mRNA | AK295685.1 | - | 1 | 123 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105371849 | LOC105371849 | -13.1kb | Anti-sense | Chr. 17 - 60079309 - 60088695 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100996660 | LOC100996660 | 23.6kb | Sense | Chr. 17 - 60126535 - 60135748 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001760.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK295685.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 269529 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 173929 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5143 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5142 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3258782 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17001878 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10399778 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979074 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979070 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17001871 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10399782 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979067 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979056 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17001908 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979030 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17001867 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979024 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979075 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979072 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979049 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979068 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17001887 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10399784 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17001865 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10399783 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17001846 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10399785 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10399777 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979040 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979031 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17001867 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10399782 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10399783 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17001878 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17001865 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979070 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979072 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979068 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979040 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17001887 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10399778 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979074 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979030 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979056 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17001908 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979075 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17001846 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17001871 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979024 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979031 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979067 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30979049 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10399777 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10399785 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10399784 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399753 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399782 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17001862 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399789 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399766 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399770 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399767 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399768 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399772 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399785 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399760 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399778 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399763 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399758 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399755 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399780 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399774 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399751 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399765 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399756 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399787 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10399775 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8017211 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8017210 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR17023500.hg.1 | NR_027408.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17023500.hg.1 | TC17001760.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011308.hg.1 | ENST00000627667.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011308.hg.1 | NR_027408.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011308.hg.1 | NR_027408.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011308.hg.1 | NR_027408.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700191123.hg.1 | NR_027408.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700191123.hg.1 | NR_027408.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700191123.hg.1 | TC1700011308.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700011308.hg.1 | ENST00000627667.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011308.hg.1 | NR_027408.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011308.hg.1 | NR_027408.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011308.hg.1 | NR_027408.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700191121.hg.1 | NR_027408.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700191121.hg.1 | NR_027408.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700191121.hg.1 | TC1700011308.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700191122.hg.1 | ENST00000627667.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700191122.hg.1 | NR_027408.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700191122.hg.1 | ENST00000627667.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700191122.hg.1 | NR_027408.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700191122.hg.1 | TC1700011308.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008653 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008653 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 189636 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1735165 | - | - |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1735165 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1735165 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1735165 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1735165 | - | - |

LOC653653
Genomic Map