Assay Details
| Gene Symbol: MIR16-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029525.1 | 1 | 35 | 81 | ||
| GenBank mRNA | AF480552.1 | - | 1 | 35 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 15b | MIR15B |
MIRN15B, hsa-mir-15b, miR-15b |
-60.0 | Sense | Chr. 3 - 160404588 - 160404685 on NCBI Build GRCh38 |
| Downstream | tripartite motif containing 59 | TRIM59 |
IFT80L, MRF1, RNF104, TRIM57, TSBF1 |
30.7kb | Anti-sense | Chr. 3 - 160435503 - 160449838 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0300009339.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000875.hg.1
Set Membership: |
|

MIR16-2
Genomic Map