Assay Details
| Gene Symbol: MIR604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030335.1 | 1 | 49 | 89 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SVIL antisense RNA 1 | SVIL-AS1 | -57.1kb | Anti-sense | Chr. 10 - 29409534 - 29487856 on NCBI Build GRCh38 | |
| Downstream | microRNA 938 | MIR938 |
MIRN938, hsa-mir-938 |
57.2kb | Sense | Chr. 10 - 29602264 - 29602346 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1000010176.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001145.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153039 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512297 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512323 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153058 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153043 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153045 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512302 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512301 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512342 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07152999 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512307 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512312 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153032 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153054 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153014 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153055 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153059 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512334 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153000 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512303 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512311 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153019 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153046 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512347 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153030 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512308 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153002 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512296 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153053 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512299 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153047 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153037 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512328 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512330 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153060 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512337 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512312 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512296 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512323 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153045 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512307 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153000 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153037 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153030 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153014 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512330 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512299 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153047 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153053 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153002 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153054 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153019 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512347 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512342 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153059 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153046 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07152999 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512308 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153055 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512302 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153039 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512297 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153043 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512311 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512328 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153060 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153032 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512301 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512337 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07153058 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512334 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27512303 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10014388.hg.1 | ENST00000384880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10014388.hg.1 | NR_030335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10014388.hg.1 | TC10001145.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010176.hg.1 | ENST00000384880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010176.hg.1 | NR_030335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000169375.hg.1 | ENST00000384880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000169375.hg.1 | NR_030335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000169375.hg.1 | ENST00000384880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000169375.hg.1 | NR_030335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000169375.hg.1 | TC1000010176.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000010176.hg.1 | ENST00000384880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010176.hg.1 | NR_030335.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3309744 | NR_030335.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3309744 | NR_030335.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309744 | NR_030335.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309744 | NR_030335.1 | human_transcriptome |

MIR604
Genomic Map