Assay Details
Gene Symbol: MIR634 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030364.1 | 1 | 46 | 87 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | PRKCA antisense RNA 1 | PRKCA-AS1 | -370.2kb | Anti-sense | Chr. 17 - 66398069 - 66416854 on NCBI Build GRCh38 | |
Downstream | calcium voltage-gated channel auxiliary subunit gamma 5 | CACNG5 | 47.9kb | Sense | Chr. 17 - 66877273 - 66885278 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700008670.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17000787.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425939 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425911 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31008321 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31008313 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425932 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425924 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425937 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425918 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425917 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425963 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425910 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31008329 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425949 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31008325 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31008283 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31008327 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425933 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31008319 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425945 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425935 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425922 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425965 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425927 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31008288 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425941 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425914 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425928 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425920 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425958 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425956 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425931 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31008297 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425947 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425913 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425959 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31008283 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425945 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425913 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425949 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31008329 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425914 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425932 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425917 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31008321 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31008319 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425928 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425941 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425920 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425958 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425911 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425937 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425927 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425956 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31008327 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425939 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31008313 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425931 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425933 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425922 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31008325 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425963 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425910 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31008288 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425947 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425918 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425935 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425965 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425959 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31008297 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10425924 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17010234.hg.1 | ENST00000385208.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17010234.hg.1 | NR_030364.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17010234.hg.1 | TC17000787.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700008670.hg.1 | ENST00000385208.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008670.hg.1 | NR_030364.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700164781.hg.1 | ENST00000385208.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700164781.hg.1 | NR_030364.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700164781.hg.1 | ENST00000385208.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700164781.hg.1 | NR_030364.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700164781.hg.1 | TC1700008670.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700008670.hg.1 | ENST00000385208.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008670.hg.1 | NR_030364.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3310633 | NR_030364.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3310633 | NR_030364.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310633 | NR_030364.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310633 | NR_030364.1 | human_transcriptome |
