Assay Details
| Gene Symbol: MIR646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030376.1 | 1 | 45 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC729296 | LOC729296 | -207.1kb | Sense | Chr. 20 - 60087686 - 60101387 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101928048 | LOC101928048 | 161.0kb | Anti-sense | Chr. 20 - 60469609 - 60472976 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC2000007950.hg.1
Set Membership: |
|

MIR646
Genomic Map