Assay Details
| Gene Symbol: MIR1205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031610.1 | 1 | 37 | 58 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 75 | TMEM75 | -11.9kb | Anti-sense | Chr. 8 - 127946559 - 127948723 on NCBI Build GRCh38 | |
| Downstream | microRNA 1206 | MIR1206 |
MIRN1206, hsa-mir-1206 |
48.2kb | Sense | Chr. 8 - 128008898 - 128008956 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0800008855.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08000752.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744358 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744326 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462262 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462231 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16134489 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744354 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462269 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462235 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462267 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462254 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462246 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462256 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462241 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744361 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744357 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744345 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744330 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462253 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462252 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744349 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462251 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744351 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462232 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462229 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462259 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462243 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744356 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462248 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462238 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462267 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744326 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16134489 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462235 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462250 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462232 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462241 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744349 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462231 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462229 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462262 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744361 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462253 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744354 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462252 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462256 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744356 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462259 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462238 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462265 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744351 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744357 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744345 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462254 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462269 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462251 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744330 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462248 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26744358 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462246 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06462243 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800008855.hg.1 | ENST00000408844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008855.hg.1 | NR_031610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08010115.hg.1 | TC08000752.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08010115.hg.1 | ENST00000408844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08010115.hg.1 | NR_031610.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800160646.hg.1 | ENST00000408844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800160646.hg.1 | NR_031610.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800160646.hg.1 | ENST00000408844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800160646.hg.1 | NR_031610.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800160646.hg.1 | TC0800008855.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800008855.hg.1 | ENST00000408844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800008855.hg.1 | NR_031610.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3310700 | NR_031610.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3310700 | NR_031610.1 | human_transcriptome |

MIR1205
Genomic Map