Assay Details
| Gene Symbol: MIR1295A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031627.1 | 1 | 43 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 1295b | MIR1295B | Anti-sense | Chr. 1 - 171101739 - 171101798 on NCBI Build GRCh38 | ||
| Downstream | flavin containing monooxygenase 6 pseudogene | FMO6P | FMO6 | 35.9kb | Anti-sense | Chr. 1 - 171137740 - 171161563 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0100016384.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01003511.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652719 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652736 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652755 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652745 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00593260 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00593280 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652751 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652721 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00593288 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00593285 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652720 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652761 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652746 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00593268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652723 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652738 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652724 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652754 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652760 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652750 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652740 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652730 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652734 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00593277 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00593272 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652726 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652758 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652722 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652727 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652737 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652724 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652737 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00593260 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652750 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652719 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652726 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652730 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652723 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00593288 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652721 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652734 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652740 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652751 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00593277 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652755 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652754 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00593285 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652746 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00593268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652760 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652761 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652727 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652720 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00593272 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652736 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652758 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652745 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00593280 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652722 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20652738 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01054563.hg.1 | TC01003511.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054563.hg.1 | ENST00000408463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054563.hg.1 | NR_031627.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016384.hg.1 | ENST00000408463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016384.hg.1 | NR_031627.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100224618.hg.1 | ENST00000408463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100224618.hg.1 | NR_031627.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100224618.hg.1 | ENST00000408463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100224618.hg.1 | NR_031627.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100224618.hg.1 | TC0100016384.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100016384.hg.1 | ENST00000408463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016384.hg.1 | NR_031627.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3309264 | NR_031627.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3309264 | NR_031627.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309264 | NR_031627.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309264 | NR_031627.1 | human_transcriptome |

MIR1295A
Genomic Map