Assay Details
| Gene Symbol: MIR1258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031659.1 | 1 | 38 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 385B | ZNF385B | ZNF533 | Sense | Chr. 2 - 179441982 - 179864550 on NCBI Build GRCh38 | |
| Downstream |
CWC22 homolog, spliceosome-associated protein |
CWC22 |
EIF4GL, NCM, fSAPb |
84.0kb | Sense | Chr. 2 - 179944876 - 180007359 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0200015096.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02002580.hg.1
Set Membership: |
|

MIR1258
Genomic Map