Assay Details
| Gene Symbol: MIR1262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031664.1 | 1 | 53 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | GNG12 antisense RNA 1 | GNG12-AS1 | Anti-sense | Chr. 1 - 67832288 - 68202987 on NCBI Build GRCh38 | ||
| Downstream |
RPE65, retinoid isomerohydrolase |
RPE65 |
BCO3, LCA2, RP20, mRPE65, rd12, sRPE65 |
245.2kb | Sense | Chr. 1 - 68428822 - 68450322 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0100014537.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002764.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297301 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297279 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297298 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00268813 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297327 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297289 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297282 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297299 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297294 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00268787 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297306 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297295 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297323 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297297 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297326 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297310 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297314 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297311 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00268788 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297286 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297280 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00268812 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297308 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00268815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297288 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00268807 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297303 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297296 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00268818 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297284 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297299 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297286 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297295 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297308 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00268788 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297289 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297314 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297301 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297294 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297288 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00268813 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297280 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00268812 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297282 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297303 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297284 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00268815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00268807 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297296 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297323 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00268787 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297306 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297327 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297298 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297311 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00268818 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297326 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297279 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297297 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20297310 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01043184.hg.1 | TC01002764.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01043184.hg.1 | ENST00000408276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01043184.hg.1 | NR_031664.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014537.hg.1 | ENST00000408276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014537.hg.1 | NR_031664.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100208580.hg.1 | ENST00000408276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100208580.hg.1 | NR_031664.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100208580.hg.1 | ENST00000408276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100208580.hg.1 | NR_031664.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100208580.hg.1 | TC0100014537.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100014537.hg.1 | ENST00000408276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014537.hg.1 | NR_031664.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3310830 | NR_031664.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3310830 | NR_031664.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310830 | NR_031664.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310830 | NR_031664.1 | human_transcriptome |

MIR1262
Genomic Map