Assay Details
| Gene Symbol: F11-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033900.1 | 2 - 3 | 490 | 131 | ||
| GenBank mRNA | BC034307.1 | - | 2 - 3 | 431 | 131 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kallikrein B1 | KLKB1 |
KLK3, PKK, PKKD, PPK |
-30.0kb | Anti-sense | Chr. 4 - 186215714 - 186258477 on NCBI Build GRCh38 |
| Downstream | melatonin receptor 1A | MTNR1A |
MEL-1A-R, MT1 |
242.0kb | Sense | Chr. 4 - 186532994 - 186555567 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04002951.hg.1
Set Membership: |
|

F11-AS1
Genomic Map