Assay Details
| Gene Symbol: LOC100129316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033912.1 | 1 - 2 | 1604 | 88 | ||
| GenBank mRNA | AK128703.1 | - | 1 - 2 | 1604 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | spleen associated tyrosine kinase | SYK | p72-Syk | -170.9kb | Anti-sense | Chr. 9 - 90801680 - 90898560 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 484 | LINC00484 | C9orf73 | 43.5kb | Anti-sense | Chr. 9 - 91119062 - 91182762 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001321.hg.1
Set Membership: |
|

LOC100129316
Genomic Map